Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VRN2
Entrez Gene ID
827392
Gene Name
VEFS-Box of polycomb protein
Gene Aliases
AT4G16845, REDUCED VERNALIZATION RESPONSE 2
Chromosome Location
Chr.4: 9476069 - 9479878 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117787.4 NP_567517.1 8 - 9 771 143
NM_202834.2 NP_974563.1 7 - 8 873 143
GenBank mRNA AY034902.1 - 8 - 9 696 143
AY063047.1 - 7 - 8 465 143

View Full Product Details