Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIP2
Entrez Gene ID
827414
Gene Name
FRIGIDA interacting protein 2
Gene Aliases
AT4G17060, FRIGIDA interacting protein 2
Chromosome Location
Chr.4: 9593381 - 9595350 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117810.7 NP_193439.1 1 - 1 1769 99

View Full Product Details