Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDG4
Entrez Gene ID
827492
Gene Name
homeodomain GLABROUS 4
Gene Aliases
AT4G17710, DL4890C, FCAALL.102, homeodomain GLABROUS 4
Chromosome Location
Chr.4: 9856255 - 9859288 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117880.3 NP_193506.2 3 - 4 436 98
GenBank mRNA AY133700.1 - 3 - 4 436 98

View Full Product Details