Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APRR2
Entrez Gene ID
827527
Gene Name
CheY-like two-component responsive regulator family protein
Gene Aliases
AT4G18020, PRR2, PSEUDO-RESPONSE REGULATOR 2, T6K21.200, T6K21_200
Chromosome Location
Chr.4: 10002846 - 10007626 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084936.2 NP_001078405.1 7 - 8 1319 132
NM_001160778.2 NP_001154250.1 7 - 8 1355 132
NM_001203830.1 NP_001190759.1 7 - 8 1342 132
NM_001341252.1 NP_001329342.1 8 - 9 1333 132
NM_001341253.1 NP_001329343.1 8 - 9 1313 132
NM_001341254.1 NP_001329339.1 7 - 8 1594 132
NM_001341255.1 NP_001329340.1 7 - 8 1574 132
NM_001341256.1 NP_001329341.1 7 - 8 1173 132
NM_117912.4 NP_567548.1 7 - 8 1355 132
NM_179072.3 NP_849403.1 7 - 8 1327 132
NM_179073.4 NP_849404.2 7 - 8 1295 132
GenBank mRNA AB041531.1 - 6 - 7 1045 132
AF385725.1 - 7 - 8 1179 132
AY143963.1 - 6 - 7 1035 132

View Full Product Details