Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APRR2
Entrez Gene ID
827527
Gene Name
CheY-like two-component responsive regulator family protein
Gene Aliases
AT4G18020, PRR2, PSEUDO-RESPONSE REGULATOR 2, T6K21.200, T6K21_200
Chromosome Location
Chr.4: 10002846 - 10007626 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084936.2 NP_001078405.1 9 - 10 1647 72
NM_001160778.2 NP_001154250.1 9 - 10 1683 72
NM_001203830.1 NP_001190759.1 9 - 10 1670 72
NM_001341252.1 NP_001329342.1 10 - 11 1661 72
NM_001341253.1 NP_001329343.1 10 - 11 1641 72
NM_001341254.1 NP_001329339.1 9 - 10 1922 72
NM_001341255.1 NP_001329340.1 9 - 10 1902 72
NM_001341256.1 NP_001329341.1 9 - 10 1501 72
NM_117912.4 NP_567548.1 9 - 10 1683 72
NM_179072.3 NP_849403.1 9 - 10 1655 72
NM_179073.4 NP_849404.2 9 - 10 1623 72
GenBank mRNA AB041531.1 - 8 - 9 1373 72
AF385725.1 - 9 - 10 1507 72
AY143963.1 - 8 - 9 1363 72

View Full Product Details