Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF4E
Entrez Gene ID
827529
Gene Name
eukaryotic translation initiation factor 4E
Gene Aliases
ARABIDOPSIS THALIANA EUKARYOTIC TRANSLATION INITATION FACTOR 4E1, AT.EIF4E1, AT4G18040, CUCUMOVIRUS MULTIPLICATION 1, CUM1, EUKARYOTIC TRANSLATION INITATION FACTOR 4E, F15J5.10, F15J5_10, eIF4E1, eukaryotic translation Initiation Factor 4E1, eukaryotic translation initiation factor 4E
Chromosome Location
Chr.4: 10016530 - 10018232 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117914.4 NP_193538.1 2 - 3 545 110
GenBank mRNA AB107250.1 - 2 - 3 497 110
AF389286.1 - 2 - 3 496 110
AY086496.1 - 2 - 3 499 110
AY093750.1 - 2 - 3 464 110
Y10548.1 - - 491 110

View Full Product Details