Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF4E
Entrez Gene ID
827529
Gene Name
eukaryotic translation initiation factor 4E
Gene Aliases
ARABIDOPSIS THALIANA EUKARYOTIC TRANSLATION INITATION FACTOR 4E1, AT.EIF4E1, AT4G18040, CUCUMOVIRUS MULTIPLICATION 1, CUM1, EUKARYOTIC TRANSLATION INITATION FACTOR 4E, F15J5.10, F15J5_10, eIF4E1, eukaryotic translation Initiation Factor 4E1, eukaryotic translation initiation factor 4E
Chromosome Location
Chr.4: 10016530 - 10018232 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117914.4 NP_193538.1 3 - 4 666 122
GenBank mRNA AB107250.1 - 3 - 4 618 122
AF389286.1 - 3 - 4 617 122
AY086496.1 - 3 - 4 620 122
AY093750.1 - 3 - 4 585 122
Y10548.1 - - 612 122

View Full Product Details