Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCB9
Entrez Gene ID
827530
Gene Name
P-glycoprotein 9
Gene Aliases
AT4G18050, ATP-binding cassette B9, F15J5.20, F15J5_20, P-glycoprotein 9, P-glycoprotein 9, PGP9
Chromosome Location
Chr.4: 10021786 - 10027425 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341257.1 NP_001328203.1 8 - 9 2772 97
NM_117915.7 NP_193539.6 8 - 9 2353 97

View Full Product Details