Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KCO6
- Entrez Gene ID
-
827541
- Gene Name
- Ca2+ activated outward rectifying K+ channel 6
- Gene Aliases
-
AT4G18160, ATKCO6, ATTPK3, CA2+ ACTIVATED OUTWARD RECTIFYING K+ CHANNEL 6, Ca2+ activated outward rectifying K+ channel 6, F15J5.130, F15J5_130, TPK3
- Chromosome Location
-
Chr.4: 10055379 - 10058185 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117926.3 |
NP_193550.1 |
1 - 2 |
1468 |
127 |
View Full Product Details