Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SS4
Entrez Gene ID
827550
Gene Name
starch synthase 4
Gene Aliases
ARABIDOPSIS THALIANA STARCH SYNTHASE 4, AT4G18240, ATSS4, SSIV, STARCH SYNTHASE 4, T9A21.90, T9A21_90, starch synthase 4
Chromosome Location
Chr.4: 10082108 - 10087203 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117934.4 NP_193558.3 2 - 3 622 116
GenBank mRNA AK226610.1 - 2 - 3 627 116
KJ676990.1 - 2 - 3 509 116

View Full Product Details