Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRH1
Entrez Gene ID
827598
Gene Name
Leucine-rich repeat protein kinase family protein
Gene Aliases
AT4G18640, F28A21.50, F28A21_50, morphogenesis of root hair 1
Chromosome Location
Chr.4: 10259600 - 10263822 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117980.5 NP_193599.3 11 - 12 2298 117
GenBank mRNA FJ708745.1 - 10 - 11 1583 117

View Full Product Details