Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTAC7
Entrez Gene ID
832500
Gene Name
plastid transcriptionally active7
Gene Aliases
AT5G24314, PDE225, PIGMENT DEFECTIVE 225, TAC7, plastid transcriptionally active7
Chromosome Location
Chr.5: 8277700 - 8279400 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203450.1 NP_001190379.1 1 - 2 91 69
NM_147910.4 NP_680215.1 1 - 2 52 69
GenBank mRNA AY063802.1 - 1 - 2 49 69
AY123012.1 - 1 - 2 41 69

View Full Product Details