Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2-A5
Entrez Gene ID
842851
Gene Name
phloem protein 2 A5
Gene Aliases
AT1G65390, AT1G65400, ATPP2-A5, T8F5.18, T8F5_18, phloem protein 2 A5
Chromosome Location
Chr.1: 24292372 - 24294740 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334203.1 NP_001319325.1 1 - 2 582 95
NM_001334204.1 NP_001322028.1 1 - 2 594 95
NM_105213.4 NP_176718.2 1 - 2 700 95
GenBank mRNA AF325110.1 - 1 - 2 506 95
AK176581.1 - 1 - 2 594 95

View Full Product Details