Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTB1
Entrez Gene ID
842855
Gene Name
global transcription factor group B1
Gene Aliases
AT1G65440, GLOBAL TRANSCRIPTION FACTOR GROUP B, T8F5.22, T8F5_22, global transcription factor group B1
Chromosome Location
Chr.1: 24306545 - 24314639 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084306.2 NP_001077775.1 6 - 7 607 117
NM_001198388.1 NP_001185317.1 7 - 8 545 117
NM_001334206.1 NP_001321759.1 6 - 7 680 117
NM_105218.5 NP_176723.3 6 - 7 626 117

View Full Product Details