Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEXO3
Entrez Gene ID
842871
Gene Name
beta-hexosaminidase 3
Gene Aliases
AT1G65590, ATHEX1, F5I14.12, F5I14_12, beta-hexosaminidase 3
Chromosome Location
Chr.1: 24385552 - 24391208 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105233.5 NP_176737.2 5 - 6 1078 127
GenBank mRNA AY128283.1 - 5 - 6 652 127
BT000831.1 - 5 - 6 634 127

View Full Product Details