Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMK1
Entrez Gene ID
842930
Gene Name
transmembrane kinase 1
Gene Aliases
AT1G66150, F15E12.4, F15E12_4, transmembrane kinase 1
Chromosome Location
Chr.1: 24631338 - 24634781 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105286.4 NP_176789.1 1 - 2 2458 86

View Full Product Details