Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFP6
Entrez Gene ID
843022
Gene Name
zinc finger protein 6
Gene Aliases
AT1G67030, F1O19.8, F1O19_8, zinc finger protein 6
Chromosome Location
Chr.1: 25016402 - 25017512 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105372.3 NP_176873.1 1 - 1 801 69
GenBank mRNA AY050387.1 - 1 - 1 660 69
AY093800.1 - 1 - 1 559 69
L39649.1 - 1 - 1 612 69

View Full Product Details