Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LBD40
Entrez Gene ID
843030
Gene Name
LOB domain-containing protein 40
Gene Aliases
AT1G67100, F1O19.17, LOB domain-containing protein 40
Chromosome Location
Chr.1: 25053673 - 25054864 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105380.5 NP_176881.1 1 - 2 321 77
GenBank mRNA AB473870.1 - 1 - 2 197 77
BT010943.1 - 1 - 2 235 77
BT011650.1 - 1 - 2 193 77

View Full Product Details