Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LINC1
- Entrez Gene ID
-
843043
- Gene Name
- little nuclei1
- Gene Aliases
-
(Japanese for nucleus) 2, AT1G67230, AtLINC1, CROWDED NUCLEI 1, CRWN1, F1N21.5, KAKU2, LITTLE NUCLEI1
- Chromosome Location
-
Chr.1: 25151211 - 25156389 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105392.5 |
NP_176892.1 |
6 - 7 |
3404 |
102 |
View Full Product Details