Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REV3
Entrez Gene ID
843071
Gene Name
recovery protein 3
Gene Aliases
AT1G67500, ATREV3, T1F15.3, T1F15_3, recovery protein 3
Chromosome Location
Chr.1: 25287356 - 25296988 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334301.1 NP_001319333.1 15 - 16 4049 74
NM_001334302.1 NP_001320390.1 16 - 17 4022 74
NM_001334303.1 NP_001320389.1 16 - 17 4003 74
NM_001334304.1 NP_001320393.1 16 - 17 3999 74
NM_001334305.1 NP_001320391.1 16 - 17 4001 74
NM_001334306.1 NP_001320392.1 15 - 16 4090 74
NM_105417.4 NP_176917.2 16 - 17 3974 74
GenBank mRNA AB114052.1 - 15 - 16 3842 74

View Full Product Details