Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSFA8
Entrez Gene ID
843125
Gene Name
heat shock transcription factor A8
Gene Aliases
AT-HSFA8, AT1G67970, T23K23.18, T23K23_18, heat shock transcription factor A8, heat shock transcription factor A8
Chromosome Location
Chr.1: 25484449 - 25486512 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105467.4 NP_176964.1 1 - 2 440 119
GenBank mRNA AJ251866.1 - 1 - 2 391 119
AY094396.1 - 1 - 2 328 119

View Full Product Details