Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIS1
Entrez Gene ID
843128
Gene Name
phosphatidylinositol synthase 1
Gene Aliases
AT1G68000, ATPIS, ATPIS1, PHOSPHATIDYLINOSITOL SYNTHASE, T23K23.15, T23K23_15, phosphatidylinositol synthase 1
Chromosome Location
Chr.1: 25490911 - 25493077 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105470.3 NP_176967.1 3 - 4 448 64
GenBank mRNA AJ000539.1 - 3 - 4 285 64
AY140047.1 - 3 - 4 340 64

View Full Product Details