Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IDD14
- Entrez Gene ID
-
843141
- Gene Name
- indeterminate(ID)-domain 14
- Gene Aliases
-
AT1G68130, AtIDD14, IDD14alpha, IDD14beta, T23K23.2, T23K23_2, indeterminate(ID)-domain 14
- Chromosome Location
-
Chr.1: 25532008 - 25534698 on Build TAIR10.1
- Gene Symbol
- AAC1
- Entrez Gene ID
-
820005
- Gene Name
- ADP/ATP carrier 1
- Gene Aliases
-
ADP/ATP carrier 1, AT3G08580
- Chromosome Location
-
Chr.3: 2605435 - 2607895 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK226814.1 |
- |
- |
1924 |
67 |
View Full Product Details