Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LINC3
Entrez Gene ID
843210
Gene Name
little nuclei3
Gene Aliases
AT1G68790, CROWDED NUCLEI 3, CRWN3, F14K14.10, F14K14_10, LITTLE NUCLEI3
Chromosome Location
Chr.1: 25834686 - 25839541 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105552.3 NP_177046.1 2 - 3 807 135

View Full Product Details