Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STN7
Entrez Gene ID
843215
Gene Name
Serine/Threonine kinase domain protein
Gene Aliases
AT1G68830, STT7 homolog STN7, T6L1.2, T6L1_2
Chromosome Location
Chr.1: 25872412 - 25875662 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105557.6 NP_564946.1 1 - 2 587 124
GenBank mRNA BT030352.1 - 1 - 2 398 124

View Full Product Details