Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SP1L2
Entrez Gene ID
843254
Gene Name
SPIRAL1-like2
Gene Aliases
AT1G69230, F4N2.18, SPIRAL1-like2
Chromosome Location
Chr.1: 26026253 - 26027771 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105590.3 NP_177083.1 2 - 3 413 62
NM_202381.2 NP_974110.1 2 - 3 344 62
GenBank mRNA BT024676.1 - 1 - 2 256 62

View Full Product Details