Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WRKY57
Entrez Gene ID
843262
Gene Name
WRKY DNA-binding protein 57
Gene Aliases
AT1G69310, ATWRKY57, F23O10.11, F23O10_11, WRKY DNA-binding protein 57
Chromosome Location
Chr.1: 26054275 - 26058383 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334406.1 NP_001321562.1 1 - 2 611 70
NM_001334407.1 NP_001321561.1 1 - 2 552 70
NM_105598.3 NP_177090.1 1 - 2 830 70
NM_202383.3 NP_974112.1 1 - 2 830 70
GenBank mRNA AB493529.1 - 1 - 2 356 70
AY071849.1 - 1 - 2 356 70
BT026057.1 - 1 - 2 356 70

View Full Product Details