Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT1G69460
Entrez Gene ID
843278
Gene Name
emp24/gp25L/p24 family/GOLD family protein
Gene Aliases
AT1G69460, F10D13.26, F10D13_26
Chromosome Location
Chr.1: 26111688 - 26113256 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105613.5 NP_177105.2 1 - 2 313 79
GenBank mRNA AY072343.1 - 1 - 2 251 79
AY114598.1 - 1 - 2 217 79

View Full Product Details