Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY36
Entrez Gene ID
843317
Gene Name
WRKY DNA-binding protein 36
Gene Aliases
AT1G69810, ATWRKY36, T17F3.16, T17F3_16, WRKY DNA-binding protein 36
Chromosome Location
Chr.1: 26276948 - 26279255 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334428.1 NP_001321291.1 3 - 4 1201 80
NM_105649.3 NP_564976.1 3 - 4 854 80
GenBank mRNA AB493530.1 - 3 - 4 715 80
AF404859.1 - 3 - 4 715 80
BT029234.1 - 3 - 4 715 80

View Full Product Details