Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NRT1:2
- Entrez Gene ID
-
843321
- Gene Name
- nitrate transporter 1:2
- Gene Aliases
-
ABA-importing transporter 1, AIT1, AT1G69850, ATNRT1:2, AtNPF4.6, NPF4.6, NRT1.2, NRT1/ PTR family 4.6, NTL1, T17F3.12, T17F3_12, nitrate transporter 1:2
- Chromosome Location
-
Chr.1: 26296540 - 26300684 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105653.5 |
NP_564978.1 |
3 - 4 |
600 |
69 |
GenBank mRNA |
BT000720.1 |
- |
3 - 4 |
355 |
69 |
View Full Product Details