Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNL4
Entrez Gene ID
843342
Gene Name
SIN3-like 4
Gene Aliases
AT1G70060, F20P5.21, F20P5_21, SIN3-like 4
Chromosome Location
Chr.1: 26383216 - 26389916 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334433.1 NP_001323064.1 6 - 7 1384 129
NM_105674.6 NP_177163.3 7 - 8 1217 129

View Full Product Details