Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH9B1
Entrez Gene ID
843408
Gene Name
glycosyl hydrolase 9B1
Gene Aliases
AT1G70710, ATGH9B1, CEL1, CELLULASE 1, F5A18.11, F5A18_11, glycosyl hydrolase 9B1
Chromosome Location
Chr.1: 26659129 - 26663337 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105739.4 NP_177228.1 1 - 2 584 70
GenBank mRNA AY048283.1 - 1 - 2 247 70
AY074552.1 - 1 - 2 209 70

View Full Product Details