Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLE17
Entrez Gene ID
843428
Gene Name
CLAVATA3/ESR-RELATED 17
Gene Aliases
AT1G70895, CLAVATA3/ESR-RELATED 17
Chromosome Location
Chr.1: 26727555 - 26729374 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105757.3 NP_565004.1 1 - 1 1096 131
GenBank mRNA AY088427.1 - 1 - 1 626 131

View Full Product Details