Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GLIP6
- Entrez Gene ID
-
843452
- Gene Name
- GDSL-motif lipase/hydrolase 6
- Gene Aliases
-
AT1G71120, F23N20.11, F23N20_11, GDSL-motif lipase/hydrolase 6
- Chromosome Location
-
Chr.1: 26821072 - 26822487 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105781.2 |
NP_177268.1 |
1 - 2 |
316 |
151 |
View Full Product Details