Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SERK1
- Entrez Gene ID
-
843513
- Gene Name
- somatic embryogenesis receptor-like kinase 1
- Gene Aliases
-
AT1G71830, ATSERK1, F14O23.21, F14O23_21, SOMATIC EMBRYOGENESIS RECEPTOR-LIKE KINASE 1, somatic embryogenesis receptor-like kinase 1
- Chromosome Location
-
Chr.1: 27018158 - 27022117 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105841.4 |
NP_177328.1 |
7 - 8 |
1108 |
76 |
GenBank mRNA |
FJ708676.1 |
- |
7 - 8 |
691 |
76 |
View Full Product Details