Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FUT13
Entrez Gene ID
843530
Gene Name
fucosyltransferase 13
Gene Aliases
ARABIDOPSIS FUCOSYLTRANSFERASE 4, AT1G71990, ATFT4, ATFUT13, F17M19.14, F17M19_14, FT4-M, FUCTC, fucosyltransferase 13
Chromosome Location
Chr.1: 27100210 - 27102292 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105857.4 NP_177344.2 2 - 3 773 97
GenBank mRNA AJ404862.1 - 2 - 3 638 97
BT026431.1 - 2 - 3 638 97
KJ138942.1 - 2 - 3 638 97

View Full Product Details