Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERO1
Entrez Gene ID
843560
Gene Name
endoplasmic reticulum oxidoreductins 1
Gene Aliases
AERO1, AT1G72280, T9N14.18, T9N14_18, endoplasmic reticulum oxidoreductins 1
Chromosome Location
Chr.1: 27211821 - 27214688 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105887.4 NP_177372.1 7 - 8 1284 113
GenBank mRNA AK227807.1 - 7 - 8 1113 113
BT008774.1 - 7 - 8 1102 113

View Full Product Details