The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198456.1 | NP_001185385.1 | 4 - 5 | 1787 | 63 |
NM_001334550.1 | NP_001320804.1 | 4 - 5 | 1808 | 63 | |
NM_105936.4 | NP_177421.1 | 4 - 5 | 1861 | 63 | |
GenBank mRNA | BT015409.1 | - | 3 - 4 | 1202 | 63 |