Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEPR1
Entrez Gene ID
843639
Gene Name
PEP1 receptor 1
Gene Aliases
AT1G73080, ATPEPR1, F3N23.28, F3N23_28, PEP1 RECEPTOR 1, PEP1 receptor 1
Chromosome Location
Chr.1: 27484240 - 27488204 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105966.2 NP_177451.1 1 - 2 3267 73
GenBank mRNA FJ708680.1 - 1 - 2 2994 73

View Full Product Details