Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIP3;1
Entrez Gene ID
843653
Gene Name
Aquaporin-like superfamily protein
Gene Aliases
ALPHA-TIP, ALPHA-TONOPLAST INTRINSIC PROTEIN, AT1G73190, T18K17.14, T18K17_14
Chromosome Location
Chr.1: 27521944 - 27523332 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105978.4 NP_177462.1 2 - 3 618 65
GenBank mRNA AY064054.1 - 2 - 3 441 65
AY117339.1 - 2 - 3 407 65

View Full Product Details