Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSL09
Entrez Gene ID
833654
Gene Name
glucan synthase-like 9
Gene Aliases
AT5G36870, ATGSL09, F5H8.14, F5H8_14, atgsl9, glucan synthase-like 9, gsl09
Chromosome Location
Chr.5: 14518008 - 14534060 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344150.1 NP_001329981.1 4 - 5 516 135
NM_001344151.1 NP_001329982.1 4 - 5 516 135
NM_001344152.1 NP_001329979.1 3 - 4 587 135
NM_001344153.1 NP_001329980.1 3 - 4 436 135
NM_123045.4 NP_198503.3 3 - 4 436 135

View Full Product Details