The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001344150.1 | NP_001329981.1 | 8 - 9 | 935 | 105 |
NM_001344151.1 | NP_001329982.1 | 8 - 9 | 935 | 105 | |
NM_001344152.1 | NP_001329979.1 | 7 - 8 | 1006 | 105 | |
NM_001344153.1 | NP_001329980.1 | 7 - 8 | 855 | 105 | |
NM_123045.4 | NP_198503.3 | 7 - 8 | 855 | 105 |