Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DEG10
Entrez Gene ID
833665
Gene Name
DegP protease 10
Gene Aliases
AT5G36950, DegP protease 10, DegP10, MLF18.70, MLF18_70, degradation of periplasmic proteins 10
Chromosome Location
Chr.5: 14594969 - 14598395 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123053.5 NP_568543.1 1 - 2 447 85
GenBank mRNA AY069888.1 - 1 - 2 435 85
BT015174.1 - 1 - 2 424 85

View Full Product Details