Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEF
Entrez Gene ID
833676
Gene Name
HIT-type Zinc finger family protein
Gene Aliases
AT5G37055, ATSWC6, SERRATED LEAVES AND EARLY FLOWERING
Chromosome Location
Chr.5: 14641442 - 14642513 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123064.3 NP_568545.1 2 - 3 331 130
GenBank mRNA AF378857.1 - 2 - 3 258 130
AK175306.1 - 2 - 3 248 130
AY050464.1 - 2 - 3 205 130

View Full Product Details