Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOS4
Entrez Gene ID
833763
Gene Name
pfkB-like carbohydrate kinase family protein
Gene Aliases
AT5G37850, ATSOS4, K18L3.1, K18L3_1, SALT OVERLY SENSITIVE 4
Chromosome Location
Chr.5: 15065300 - 15069094 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085208.2 NP_001078677.1 2 - 3 309 96
NM_001203503.1 NP_001190432.1 2 - 3 202 96
NM_001344222.1 NP_001331291.1 1 - 2 401 96
NM_123144.4 NP_198601.2 2 - 3 276 96
GenBank mRNA AF404865.2 - 1 - 2 80 96
AY084419.1 - 1 - 2 110 96
AY136333.1 - 2 - 3 168 96
BT008815.1 - 1 - 2 80 96

View Full Product Details