Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HA8
Entrez Gene ID
823281
Gene Name
H[+]-ATPase 8
Gene Aliases
AHA8, AT3G42640, H(+)-ATPase 8
Chromosome Location
Chr.3: 14724103 - 14728327 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114131.3 NP_189850.1 9 - 10 2606 88

View Full Product Details