Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE38
Entrez Gene ID
823389
Gene Name
maternal effect embryo arrest 38
Gene Aliases
AT3G43160, maternal effect embryo arrest 38
Chromosome Location
Chr.3: 15166561 - 15168225 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114184.1 NP_189902.1 2 - 3 164 61

View Full Product Details