Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATMIN7
Entrez Gene ID
823405
Gene Name
HOPM interactor 7
Gene Aliases
AT3G43300, BEN1, BFA-VISUALIZED ENDOCYTIC TRAFFICKING DEFECTIVE1, HOPM interactor 7, MIN7
Chromosome Location
Chr.3: 15233845 - 15245281 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203077.1 NP_001190006.1 29 - 30 4721 149
NM_001339097.1 NP_001327198.1 29 - 30 4905 149
NM_114198.4 NP_189916.4 29 - 30 4715 149

View Full Product Details