Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JAZ11
Entrez Gene ID
823426
Gene Name
jasmonate-zim-domain protein 11
Gene Aliases
AT3G43440, TIFY3A, jasmonate-zim-domain protein 11
Chromosome Location
Chr.3: 15367546 - 15370157 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114212.5 NP_189930.1 3 - 4 419 66
GenBank mRNA BT015154.1 - 3 - 4 309 66
BT015661.1 - 3 - 4 295 66

View Full Product Details