The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001339113.1 | NP_001327805.1 | 6 - 7 | 1210 | 63 |
NM_114254.3 | NP_189972.2 | 6 - 7 | 1210 | 63 | |
GenBank mRNA | AY072099.1 | - | 6 - 7 | 1170 | 63 |